Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ammonium Carbonate, 10% w/v, Reagents
Click to view available options
Quantity:
10 L
20 L
Specifications
Specifications
Boiling Point | 100°C |
Color | Colorless |
Physical Form | Liquid |
Quantity | 10 L |
Percent Purity | 10% w/v |
Grade | Reagent |
Packaging | Cubitainer |
Specific Gravity | 1.05 |
Chemical Name or Material | Ammonium Carbonate, 10% w/v |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction