Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium carbonate, Puratronic™, 99.999% (metals basis)

Catalog No. AA1298030
Click to view available options
Quantity:
10 g
50 g
250 g

CAS: 506-87-6 | CH8N2O3 | 96.086 g/mol

Ammonium carbonate is widely utilized as a leavening agent in lebkuchen, cookies and flat biscuits. It finds an important application as an emetic and an active ingredient in some cough syrups. It is the main component in smelling salts and an active ingredient in some smokeless tobacco products, shampoos and dyes used in textile industries. It is an important predecessor to the modern leavening agent’s baking soda and baking powder. It serves as a foaming agent for the production of expanded material. It is also used as a reagent for analytical purposes in the chemical industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 506-87-6
MDL Number MFCD00010890

Specifications

Melting Point 60° (sublimation)
Physical Form Powder
Quantity 250 g
Assay Percent Range 99.999% (metals basis)
Molecular Formula CH8N2O3
Beilstein 3627235
Synonym ammonium carbonate, diammonium carbonate, carbonic acid, diammonium salt, ammonia sesquicarbonate, carbonic acid, ammonium salt, caswell no. 042, ammoniumcarbonat german, unii-pdp691cn28, carbonic acid, ammonium salt 1:2, ammoniumcarbonate
Solubility Information Soluble in water.
InChI Key PRKQVKDSMLBJBJ-UHFFFAOYSA-N
SMILES C(=O)([O-])[O-].[NH4+].[NH4+]
IUPAC Name diazanium;carbonate
Molecular Weight (g/mol) 96.086
PubChem CID 517111
Formula Weight 96.09
Sensitivity Hygroscopic
Chemical Name or Material Ammonium Carbonate
Show More Show Less
Hazard Category H302
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 208-058-0
RTECSNumber BP1925000
TSCA Yes
storageNote1 Hygroscopic
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.