Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium carbonate, ACS, NH{3} 30.0+%

Catalog No. aa3622936
Click to view available options
Quantity:
500 g
2 kg

In baking powder, smelling salts, as an analytical lab reagent, manufacturing of rubber materials, casein glue | CAS: 506-87-6 | CH8N2O3 | 96.086 g/mol

In baking powder, smelling salts, as an analytical lab reagent, manufacturing of rubber materials, casein glue, and fire extinguishing compoundsAmmonium carbonate is widely utilized as a leavening agent in lebkuchen, cookies and flat biscuits. It finds an important application as an emetic and an active ingredient

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • In baking powder, smelling salts, as an analytical lab reagent, manufacturing of rubber materials, casein glue, and fire extinguishing compounds
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 506-87-6
MDL Number MFCD00010890

Specifications

Melting Point 60° (sublimation)
Physical Form Powder
Quantity 500 g
Assay Percent Range NH3 30.0+%
Molecular Formula C2H11N3O5
Beilstein 3627235
Merck Index 14,508
Synonym ammonium carbonate nf, ugam, ammonium bicarbonate-ammonium carbamate, ammonium carbonate chips nf, ammonium bicarbonate, mixt. with ammonium carbamate, ammonium carbamate, mixt. with ammonium bicarbonate, monoammonium carbonate mixture with ammonium carbamate, carbamic acid, monoammonium salt, mixt. with ammonium carbonate, diazanium hydrogen carbonate carbamate
Solubility Information Slowly soluble in water (1:4 portion); Decomposes in hot water.
InChI Key YNSUIDWLZCFLML-UHFFFAOYSA-N
SMILES C(=O)(N)[O-].C(=O)(O)[O-].[NH4+].[NH4+]
IUPAC Name diazanium;hydrogen carbonate;carbamate
Molecular Weight (g/mol) 157.126
PubChem CID 24660
Formula Weight 96.09
Grade ACS
Sensitivity Hygroscopic
Chemical Name or Material Ammonium carbonate
Show More Show Less
Hazard Category H302
Hazard Statement GHS H Statement
H302-H315-H319-H335
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 208-058-0
RTECSNumber BP1925000
TSCA Yes
storageNote1 Hygroscopic
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.