Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium cerium(IV) nitrate, 99%, for analysis

Catalog No. AC201440050
Change view
Click to view available options
Quantity:
100 g
250 g
1 kg
5 kg
Packaging:
Glass bottle
Plastic bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC201440050 5 kg Plastic bottle
AC201441000 100 g Glass bottle
AC201442500 250 g Glass bottle
AC201440010 1 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC201440050 Supplier Thermo Scientific Chemicals Supplier No. 201440050
Add to Cart

CAS: 16774-21-3 | H8CeN8O18 | 548.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent for analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16774-21-3
Molecular Formula H8CeN8O18
Molecular Weight (g/mol) 548.22
MDL Number MFCD00151121
Synonym Ceric ammonium nitrate, CAN

Specifications

Melting Point 107.0°C to 108.0°C
Color Colorless to Yellow
Physical Form Fuming Liquid
Quantity 5 kg
Assay Percent Range 99%
Linear Formula (NH4)2Ce(NO3)6
Fieser 01,120; 02,63; 03,44; 04,71; 05,101; 06,99; 09,99; 10,79; 15,70; 16,66; 17,68
Merck Index 15,1991
Solubility Information (10% in water) Clear
Formula Weight 548.22
Percent Purity 99%
Grade Analytical
Packaging Plastic bottle
Density 1.05
Chemical Name or Material Ammonium cerium(IV) nitrate, p.a.
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May intensify fire; oxidizer.
May be corrosive to metals.
Harmful if swallowed.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately
EINECSNumber 240-827-6
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.