Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Cerium(IV) Nitrate 98.0+%, TCI America™
SDP

Catalog No. C1806500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
50 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C1806500G 500 g
C180650G 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C1806500G Supplier TCI America Supplier No. C1806500G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 16774-21-3
Molecular Formula H8CeN8O18
MDL Number MFCD00151121
Synonym Ceric Ammonium Nitrate, CAN, Cerium(IV) Diammonium Nitrate, Diammonium Cerium(IV) Nitrate

Specifications

Color Red-Yellow
Physical Form Crystalline Powder
Quantity 500 g
UN Number 1477
Formula Weight 548.22
Percent Purity ≥98.0% (T)
Chemical Name or Material Ammonium Cerium(IV) Nitrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.