Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Chloride, 1.15 g/L, AquaPhoenix Scientific™
SCI_ED

Catalog No. S88485 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S88485 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S88485 Supplier Aqua Phoenix Scientific Supplier No. AC8200Q
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Laboratory grade ammonium chloride solution for analytical testing applications.

Chemical Identifiers

CAS 12125-02-9 , 7732-18-5
Molecular Formula ClH4N
Molecular Weight (g/mol) 53.49
MDL Number MFCD00011420
InChI Key NLXLAEXVIDQMFP-UHFFFAOYSA-N
IUPAC Name amine hydrochloride
SMILES N.Cl

Specifications

CAS Max % <0.01%, >99%
Physical Form Liquid
Quantity 1 L
Concentration or Composition (by Analyte or Components) 1.15 g/L
Packaging Poly Bottle
Chemical Name or Material Ammonium Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.