Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Ammonium Chloride
SCI_ED

Catalog No. S25168 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 g
500 g
2 kg
Packaging:
Lab Grade
Reagent Grade
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25168 100 g Reagent Grade
S25168A 500 g Reagent Grade
S25168C 2 kg Lab Grade
S25168B 500 g Lab Grade
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. S25168
Freezing point depression

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 12125-02-9
Molecular Formula ClH4N
Molecular Weight (g/mol) 53.49
MDL Number MFCD00011420
InChI Key NLXLAEXVIDQMFP-UHFFFAOYSA-N
Synonym ammonium chloride, salmiac, sal ammoniac, ammonium muriate, ammoniumchlorid, ammoneric, darammon, amchlor, ammonium chloride nh4 cl, ammoniumklorid
PubChem CID 25517
ChEBI CHEBI:31206
IUPAC Name amine hydrochloride
SMILES N.Cl

Specifications

Color White
Quantity 100 g
Assay Percent Range 100% w/v
Linear Formula NH4Cl
Formula Weight 53.49
Grade Laboratory
Packaging Reagent Grade
Chemical Name or Material Ammonium Chloride
Safety handling Check Alert storage code Red
ShelfLife 5 years
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.