Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ammonium Chloride, 2% (w/v), Ricca Chemical
Shop All Ricca Chemical Company Products

Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Natural Poly Bottle
Specifications
Specifications
| Color | Colorless |
| CAS Min % | 96.0, 1.0 |
| CAS Max % | 100.0, 3.0 |
| Percent Purity | 2% |
| Grade | Laboratory |
| Packaging | Natural Poly Bottle |
| Solvent or Matrix | Aqueous Solution |
| Quantity | 4 L |
| Physical Form | Liquid |
| Concentration or Composition Notes | 2% (w/v) |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction