Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Chloride, 8.5% (w/v) , with Brij™35, Ricca Chemical
SDP

Catalog No. 625216 Shop All Ricca Chemical Company Products
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Natural Poly Bottle

Chemical Identifiers

CAS 12125-02-9, 7732-18-5, 9002-92-0
Molecular Formula ClH4N
Molecular Weight (g/mol) 53.49
MDL Number MFCD00011420
InChI Key NLXLAEXVIDQMFP-UHFFFAOYSA-N
Synonym ammonium chloride, salmiac, sal ammoniac, ammonium muriate, ammoniumchlorid, ammoneric, darammon, amchlor, ammonium chloride nh4 cl, ammoniumklorid
PubChem CID 25517
ChEBI CHEBI:31206
IUPAC Name amine hydrochloride
SMILES N.Cl

Specifications

Color Colorless
CAS Min % 8.0, 90.0, 0.05
CAS Max % 9.0, 94.0, 0.05
Percent Purity 8.5%
Grade Laboratory
Packaging Natural Poly Bottle
Solvent or Matrix Aqueous Solution
Quantity 500 mL
Physical Form Liquid
Concentration or Composition Notes 8.5% (w/v)
Chemical Name or Material Ammonium Chloride, With Brij 35
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.