Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium chloride, 99.5% min

Catalog No. p-3256878
Click to view available options
Quantity:
500 g
2 kg

CAS: 12125-02-9 | ClH4N | 53.49 g/mol

It act as a buffer, as a flux in galvanizing steel sheet. It is used in batteries, to clean soldering irons, analytical lab reagent, in washing powders, electroplating, lustering cotton.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12125-02-9
Molecular Formula ClH4N
Molecular Weight (g/mol) 53.49
MDL Number MFCD00011420
InChI Key NLXLAEXVIDQMFP-UHFFFAOYSA-N
Synonym ammonium chloride, salmiac, sal ammoniac, ammonium muriate, ammoniumchlorid, ammoneric, darammon, amchlor, ammonium chloride nh4 cl, ammoniumklorid
PubChem CID 25517
ChEBI CHEBI:31206
SMILES N.Cl

Specifications

Melting Point 340° (sublimation)
Physical Form Granules
Quantity 500 g
Assay Percent Range 99.5% min.
Solubility Information Soluble in liquid ammonia,acetone,hydrazine,alcohol. Insoluble in diethyl ether,ethyl acetate.
IUPAC Name amine hydrochloride
Formula Weight 53.49
Odor Odorless
Refractive Index 1.642
Sensitivity Hygroscopic
Appearance Granular
Density 1.53 g/cm3
Chemical Name or Material Ammonium chloride
Show More Show Less
Hazard Category H302-H319-H335-H373
Precautionary Statement P260-P264b-P270-P271-P280i-P301+P312-P304+P340-P305+P351+P338-P312-P314-P330-P501c
EINECSNumber 235-186-4
RTECSNumber BP4550000
TSCA Yes
storageNote1 Hygroscopic
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.