Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Chloride, ACS Reagent, Reagents

Catalog No. 01338104
Change view
Click to view available options
Quantity:
500 g
3 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-338-104 500 g
01-338-105 3 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-338-104 Supplier Reagents Holdings Llc Supplier No. C1132000500A5
Add to Cart
Add to Cart

Ammonium Chloride, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 12125-02-9
Molecular Formula ClH4N
Molecular Weight (g/mol) 53.49
MDL Number MFCD00011420
InChI Key NLXLAEXVIDQMFP-UHFFFAOYSA-N
Synonym Ammonium Muriate
IUPAC Name amine hydrochloride
SMILES N.Cl

Specifications

Boiling Point 520°C
Melting Point 337.8°C
Color White
Physical Form Solid
Quantity 500 g
Grade ACS
Packaging Poly Bottle
Specific Gravity 1.5274
Chemical Name or Material Ammonium Chloride, ACS Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.