Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ammonium Chloride-Ammonium Hydroxide, Buffer Solution, pH 10.0 ±0.1, For Water Hardness, APHA, Spectrum™ Chemical

Click to view available options
Quantity:
500 mL
1 L
4 L
Description
Spectrum™ Chemical Ammonium Chloride-Ammonium Hydroxide, Buffer Solution, pH 10.0 ± 0.1, For Water Hardness, APHA is an aqueous inorganic buffer solution concentrate that, when diluted, is used for the American Public Health Association's (APHA) method for determination of water hardness. Find the high quality buffer solution Ammonium Chloride-Ammonium Hydroxide at Spectrum. As an aqueous inorganic buffer solution concentrate, it is used for the American Public Health Association's (APHA) method for determination of water hardness after dilution.
Specifications
Specifications
Chemical Name or Material | Ammonium Chloride-Ammonium Hydroxide |
CAS | 1336-21-6, 12125-02-9, 7732-18-5 |
CAS Max % | 1%, 98% |
UN Number | UN2672 |
Grade | APHA/EPA |
Packaging | Amber Glass Bottle |
pH | 10 ±0.1 |
Quantity | 500 mL |
Physical Form | Chemical Solution |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction