Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ammonium Chloride/ EDTA Buffer, pH 8.5, Reagents
Click to view available options
Quantity:
1 L
4 L
10 L
20 L
Specifications
Specifications
Chemical Name or Material | Ammonium Chloride/ EDTA Buffer, pH 8.5 |
Boiling Point | 100°C |
Color | Colorless |
CAS | 7732-18-5, 12125-02-9, 12125-02-9 |
pH | 8.5 |
Physical Form | Liquid |
Quantity | 1 L |
Grade | Reagent |
Packaging | Poly Bottle |
Specific Gravity | 1.005 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction