Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ammonium Chloride, Granular, Reagent, ACS, 99.5%, Spectrum™ Chemical
Shop All Spectrum Chemical Manufacturing Corporation Products

Click to view available options
Quantity:
125 g
500 g
2.5 kg
6 x 500 g
12 kg
4 x 2.5 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
Description
Spectrum™ Chemical Ammonium Chloride, Granular, Reagent, ACS is an inorganic compound, that presents itself as a white crystalline salt that is highly soluble in water. It occurs from the reaction of hydrochloric acid and ammonia. It has uses in medicine as an expectorant in cough medicine, in metalwork as a flux in preparing metals to be tin coated as well as in laboratories to produce cooling baths at low temperatures. As an ACS grade Reagent, Spectrum Chemical manufactured Ammonium Chloride is used as the quality standard against which other substances are graded and has met the toughest regulatory standards for quality and pureness.
Specifications
Specifications
pH | 4.5-5.5 |
CAS Max % | 100% |
Quantity | 12 kg |
Percent Purity | 99.5% |
Grade | ACS |
Packaging | Poly Pail |
Residue after Ignition | 0.01% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction