Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Chloride / Hydroxide, 4 Molar, Reagents

Catalog No. 01336045
Click to view available options
Quantity:
1 L
20 L

Ammonium Chloride / Hydroxide, 4 Molar, Reagent, use for EDTA titrations. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5, 12125-02-9, 12125-02-9
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
IUPAC Name water
SMILES O

Specifications

Boiling Point 100°C
Color Colorless
For Use With (Application) Titration
pH 10
Physical Form Liquid
Quantity 1 L
Solution Type Buffer
Packaging Poly Bottle
Specific Gravity 1.04
Sterility Non-sterile
Grade Reagent
Chemical Name or Material Ammonium Chloride / Hydroxide, 4 Molar
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.