Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Citrate Solution, USP <251> for Lead, Ricca Chemical
SDP

Catalog No. 6279216 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Natural Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
62792-16 500 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 62792-16 Supplier Ricca Chemical Company Supplier No. 627.9216
Add to Cart
Add to Cart

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

missing translation for 'nonReturnableMessage' missing translation for 'returnPolicyLabel'

Chemical Identifiers

CAS 7732-18-5 , 77-92-9 , 1336-21-6
Synonym dihydrogen oxide, dihydrogen monoxide

Specifications

CAS Min % 0.54, 0.2214, 0.2143
CAS Max % 0.57, 0.2304, 0.2231
Color Colorless
Boiling Point Approximately 100°C
Packaging Natural Poly Bottle
Quantity 500 mL
Grade Laboratory
Physical Form Liquid
Chemical Name or Material Ammonium Citrate Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.