Alfa Aesar™ Ammonium cobalt(II) sulfate hexahydrate, 99+%

As spectroscopic standard, cobalt plating, catalysis, ceramics

Manufacturer: Alfa Aesar™ 4390236

Quantity 500g

 View more versions of this product

Catalog No. AA4390236

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 13586-38-4
    Molecular Formula H8CoN2O8S2·6H2O
    Linear Formula (NH4)2Co(SO4)2·6H2O
    Formula Weight 395.22 (287.14 Anhydrous)
    MDL Number MFCD00151049
    UN Number UN3077
    Synonym Cobalt(II) ammonium sulfate hexhydrate; Cobaltous ammonium sulfate hexahydrate
    Chemical Name or Material Ammonium cobalt (II) sulfate hexahydrate

    Specifications

    Formula Weight 395.22 (287.14 Anhydrous)
    Health Hazard 3 GHS P Statement
    P261-P280-P301+P312-P363-P321-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Wash contaminated clothing before reuse.
    Specific treatment (see label).
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Odor Odorless
    CAS 13586-38-4
    Physical Form Crystalline
    Melting Point ≈120°C (dec.)
    Health Hazard 2 GHS H Statement
    H302-H317
    Harmful if swallowed.
    May cause an allergic skin reaction.
    MDL Number MFCD00151049
    Color Pink to red
    Quantity 500g
    Density 1.902g/mL
    TSCA TSCA
    Sensitivity Hygroscopic
    Linear Formula (NH4)2Co(SO4)2·6H2O
    Synonym Cobalt(II) ammonium sulfate hexhydrate; Cobaltous ammonium sulfate hexahydrate
    Solubility Information Soluble in water, Insoluble in alcohol
    Health Hazard 1 Warning
    DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
    Molecular Formula H8CoN2O8S2·6H2O
    UN Number UN3077
    Chemical Name or Material Ammonium cobalt (II) sulfate hexahydrate
    Assay ≥99%

    GHS H Statement
    H302-H317
    Harmful if swallowed.
    May cause an allergic skin reaction.

    GHS P Statement
    P261-P280-P301+P312-P363-P321-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Wash contaminated clothing before reuse.
    Specific treatment (see label).
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit