Learn More
Ammonium diamminetetrathiocyanatochromate(III) monohydrate, ACS, 93.0% min
Description
Ammonium diamminetetrathiocyanatochromate(III) monohydrate is used to precipitate primary and secondary amines as their ammonium salts. It is a color reagent and precipitant for mercury. It is also used as a precipitant for primary and secondary amines and certain amino acids. It is a reagent used for quantitative studies, including titrimetric, gravimetric, and spechtometric assays. Additionally, Reinecke salt has also been shown to determine the concentration of Dimenhydrinate using a spectrophotometric procedure. Other studies use Reinecke salt as a chemical actinometer to calculate the photodegradation of Manidipine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Color | Red |
Physical Form | Powder |
Quantity | 25 g |
Linear Formula | NH4[Cr(NH3)2(SCN)4]·H2O |
UN Number | UN3077 |
Merck Index | 14,8128 |
Solubility Information | Insoluble in dilute hydrochloric acid; Moderately soluble in water; Soluble in alcohol |
Formula Weight | 354.44 (336.42 Anhydrous) |
Percent Purity | ≥93.0% |
Grade | ACS |
Show More |
Safety and Handling
Hazard Category | H302+H312+H332 |
Hazard Statement | GHS H Statement H302-H312-H332 Harmful if swallowed. Harmful in contact with skin. Harmful if inhaled. |
Precautionary Statement | P260-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P314-P330-P332+P313-P362-P501c |
DOTInformation | Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S. |
EINECSNumber | 237-003-3 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.