Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium dichromate, ACS reagent

Catalog No. ac446921000
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle

CAS: 7789-09-5 | H8Cr2N2O7 | 252.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-09-5
Molecular Formula H8Cr2N2O7
Molecular Weight (g/mol) 252.07
InChI Key JOSWYUNQBRPBDN-UHFFFAOYSA-P
Synonym ammonium dichromate, ammonium bichromate, diammonium dichromate, ammonium dichromate vi, ammoniumbichromaat dutch, ammoniumdichromaat dutch, ammoniumdichromat german, unii-5j18bp595g, hsdb 481
PubChem CID 24600
IUPAC Name diazanium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium
SMILES [NH4+].[NH4+].[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-]

Specifications

Melting Point 170.0°C
Quantity 100 g
Linear Formula (NH4)2Cr2O7
Merck Index 15,512
Solubility Information Solubility in water: 360g/L (20°C). Other solubilities: soluble in alcohol
Formula Weight 252.07
Grade ACS Reagent
Packaging Plastic bottle
Chemical Name or Material Ammonium dichromate
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful in contact with skin.
Very toxic to aquatic life with long lasting effects.
May damage fertility.
May damage the unborn child.
May cause genetic defects.
Causes damage to organs through prolonge
EINECSNumber 232-143-1

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.