Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium fluoride, 98+%, extra pure

Catalog No. p-45471
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic Drum
Plastic bottle

CAS: 12125-01-8 | FH4N | 37.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12125-01-8
Molecular Formula FH4N
Molecular Weight (g/mol) 37.04
MDL Number MFCD00011423
InChI Key LDDQLRUQCUTJBB-UHFFFAOYSA-N
IUPAC Name amine hydrofluoride
SMILES N.F

Specifications

Color Colorless or White
Physical Form Adhering Crystals and/or Chunks
Quantity 100 g
Assay Percent Range 98+%
Linear Formula NH4F
Merck Index 15, 519
Solubility Information Solubility in water: soluble. Other solubilities: slightly soluble in alcohol
Formula Weight 37.04
Percent Purity ≥98%
Grade Extra Pure
Packaging Plastic bottle
Residue after Ignition 0.01 % max. (SO4)
Chemical Name or Material Ammonium fluoride, p.a.
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic in contact with skin.
Toxic if inhaled.
Toxic if swallowed.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air
EINECSNumber 235-185-9
RTECSNumber BQ6300000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.