Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ammonium fluoride, ACS, 98.0% min
Click to view available options
Quantity:
25 g
250 g
1 kg
5 x 1 kg
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Specifications
Specifications
| Color | White |
| Physical Form | Crystalline |
| Quantity | 250 g |
| Assay Percent Range | 98% min. |
| Solubility Information | Freely soluble in water. Slightly soluble in alcohol. |
| Formula Weight | 37.04 |
| Grade | ACS |
| Sensitivity | Hygroscopic |
| Appearance | White crystalline |
| Density | 1.01 g/cm3 |
| Show More |
Safety and Handling
| Hazard Category | H301+H311+H331 |
| Precautionary Statement | P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P311-P312-P330-P361-P363-P501c |
| DOTInformation | DOT Class: 6.1, Packaging Group: III |
| EINECSNumber | 235-185-9 |
| RTECSNumber | BQ6300000 |
| TSCA | Yes |
| storageNote1 | Hygroscopic |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction