Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ammonium Formate in Water, 10mM, with 0.05% Formic Acid, LC-MS
Shop All Fisherbrand Products
Click to view available options
Quantity:
1 L
Packaging:
Borosilicate Glass Bottle
Description
Ammonium Formate in Water, 10mM, with 0.05% Formic Acid, 0.57 to 0.69g/L (Ammonium Formate, w/v), CAS Number-7732-18-5, 540-69-2, 64-18-6, dihydrogen oxide, dihydrogen monoxide, 1L, <-0.005 AU at 254nm, <-1.0 AU at 220nm, Pass Test (LC-UV Gradient Suitability), <-5ppb, 100 deg.C, <-25ppb
- Suitable for use in LC/MS and UHPLC applications
- Pre-mixed for ease-of-use
- Mobile phase blend suitable for vitamin D and drugs of abuse testing
- Packaged in borosilicate glass bottles to prevent metal adduct formation, which can be used directly on the instrument.
- Ammonium formate, a commonly used HPLC buffer that is compatible with LC/MS
Specifications
Specifications
| Boiling Point | 100°C |
| Chemical Name or Material | Ammonium Formate in Water, 10mM, with 0.05% Formic Acid |
| Name Note | Mobile-phase solvent blend |
| CAS | 7732-18-5, 540-69-2, 64-18-6 |
| pH | 3.5 |
| Physical Form | Liquid |
| Quantity | 1 L |
| Synonym | dihydrogen oxide, dihydrogen monoxide |
| Grade | UHPLC-MS |
| Concentration or Composition (by Analyte or Components) | 10mM Ammonium Formate |
| Show More |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction