Learn More
Ammonium hydrogen carbonate, 98%
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Ammonium bicarbonate is a commonly used reagent for industrial and research procedures. It acts a good buffer in lyophilization and matrix assisted laser desorption. It is also utilized for the in-gel digestion of proteins by trypsin and in the MALDI mass spectrometric analysis of proteins. It is also used to make other ammonium compounds, in food processing, and for other uses. Ammonium bicarbonate can be used to study biological buffers. It is also used in a study that demonstrated that ammonium bicarbonate salts, which can be regenerated using low-temperature waste heat, can also produce sufficient voltage for hydrogen gas generation in a microbial reverse-electrodialysis electrolysis cells. It has also been used in a study that developed a fast and sensitive method for the simultaneous determination of Sudan dyes in food samples using partial filling micellar electrokinectic chromatography-mass spectrometry.
Notes
Store away from oxidizing agents, acids and bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Specifications
| Melting Point | ∼60°C (decomposition) |
| Quantity | 2500 g |
| Beilstein | 4329606 |
| Merck Index | 14,497 |
| Solubility Information | Soluble in water (220mg/ml at 20°C). Insoluble in alcohol,ethanol,benzene,and acetone. |
| Formula Weight | 79.06 |
| Percent Purity | 98% |
| Odor | Like Ammonia |
| Density | 1.586 |
| Chemical Name or Material | Ammonium hydrogen carbonate |
Safety and Handling
| Hazard Category | H302 |
| Hazard Statement | GHS H Statement H302 Harmful if swallowed. |
| Precautionary Statement | P264b-P270-P301+P312-P330-P501c |
| EINECSNumber | 213-911-5 |
| RTECSNumber | BO8600000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.