Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Ammonium hydrogen carbonate, 98%

Click to view available options
Quantity:
500g
2500g
1066-33-7
CH5NO3
79.055
MFCD00012138
ATRRKUHOCOJYRX-UHFFFAOYSA-N
ammonium bicarbonate,ammonium hydrogencarbonate,ammonium hydrogen carbonate,carbonic acid, monoammonium salt,monoammonium carbonate,ammonium acid carbonate,ammonium hydrogencarbonat,ammoniumbicarbonate,acid ammonium carbonate,ccris 7327
14013
azanium;hydrogen carbonate
C(=O)(O)[O-].[NH4+]
1066-33-7
CH5NO3
79.055
MFCD00012138
ATRRKUHOCOJYRX-UHFFFAOYSA-N
ammonium bicarbonate,ammonium hydrogencarbonate,ammonium hydrogen carbonate,carbonic acid, monoammonium salt,monoammonium carbonate,ammonium acid carbonate,ammonium hydrogencarbonat,ammoniumbicarbonate,acid ammonium carbonate,ccris 7327
14013
azanium;hydrogen carbonate
C(=O)(O)[O-].[NH4+]
Applications
Ammonium bicarbonate is a commonly used reagent for industrial and research procedures. It acts a good buffer in lyophilization and matrix assisted laser desorption. It is also utilized for the in-gel digestion of proteins by trypsin and in the MALDI mass spectrometric analysis of proteins. It is also used to make other ammonium compounds, in food processing, and for other uses. Ammonium bicarbonate can be used to study biological buffers. It is also used in a study that demonstrated that ammonium bicarbonate salts, which can be regenerated using low-temperature waste heat, can also produce sufficient voltage for hydrogen gas generation in a microbial reverse-electrodialysis electrolysis cells. It has also been used in a study that developed a fast and sensitive method for the simultaneous determination of Sudan dyes in food samples using partial filling micellar electrokinectic chromatography-mass spectrometry.

Notes
Store away from oxidizing agents, acids and bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Beilstein 4329606
Merck Index 14,497
Solubility Information Soluble in water (220mg/ml at 20°C). Insoluble in alcohol,ethanol,benzene,and acetone.
Formula Weight 79.06
Percent Purity 98%
Density 1.586
Odor Like Ammonia
Melting Point ~60°C (decomposition)
Quantity 500g
Chemical Name or Material Ammonium hydrogen carbonate

GHS H Statement
H302
Harmful if swallowed.

P264b-P270-P301+P312-P330-P501c

H302

EINECSNumber : 213-911-5

RTECSNumber : BO8600000

TSCA : Yes

Recommended Storage : Ambient temperatures