Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium hydrogen carbonate, 99.0%

Catalog No. AA14249A7
Click to view available options
Quantity:
1 kg
5 kg

CAS: 1066-33-7 | C3H8N2O9Zn2 | 346.86 g/mol

In cooling baths, fire extinguishing compounds, baking powder formulations, and in the manufacture of ceramics and pigments. Also used for compost treatment to accelerate decomposition.Ammonium hydrogen carbonate is used as a reagent in chemical industries. It is an active component used in the manufacturing of fire-extinguishing compounds, pharmaceuticals, dyes, pigments and fertilizers. It finds application in the plastic, ceramics, leather and rubber industries. It is also used in cooling baths, scale and gypsum removing agents, leavening agent for baked goods and a defatting agent for textiles. Further, it acts as a buffer used in the purification of chemicals. In addition to this, it is involved in the gel digestion of proteins by trypsin and mass spectrometric analysis of proteins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • In cooling baths, fire extinguishing compounds, baking powder formulations, and in the manufacture of ceramics and pigments
  • Also used for compost treatment to accelerate decomposition
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1066-33-7
Molecular Formula CH5NO3
Molecular Weight (g/mol) 79.06
MDL Number MFCD00012138
InChI Key ATRRKUHOCOJYRX-UHFFFAOYSA-N
Synonym ammonium bicarbonate, ammonium hydrogencarbonate, ammonium hydrogen carbonate, carbonic acid, monoammonium salt, monoammonium carbonate, ammonium acid carbonate, ammonium hydrogencarbonat, ammoniumbicarbonate, acid ammonium carbonate, ccris 7327
PubChem CID 14013
IUPAC Name carbonic acid amine
SMILES N.OC(O)=O

Specifications

Melting Point ∼60°C (decomposition)
Color White
Physical Form Powder
Quantity 5 kg
Linear Formula (NH4)HCO3
Beilstein 4329606
Merck Index 14,497
Solubility Information Soluble in cold water but decomposed by hot water; Insoluble in alcohol and acetone.
Formula Weight 79.06
Percent Purity 99.0%
Odor Like ammonia
Density 1.586 g/mL
Chemical Name or Material Ammonium hydrogen carbonate
Show More Show Less
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 213-911-5
RTECSNumber BO8600000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.