Alfa Aesar™ Ammonium hydrogen phosphate, tech., P2O5 53% min

Corrosion inhibitors, in fermentation, as flux for soldering tin, copper, brass, and zinc

Manufacturer: Alfa Aesar™ 10301A1

Quantity 1kg

 View more versions of this product

Catalog No. AA10301A1

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7783-28-0
    Formula Weight 132.06
    MDL Number MFCD00010891
    Merck Index 14,542
    Synonym Diammonium hydrogen phosphate; Phosphoric acid diammonium salt
    Chemical Name or Material Ammonium hydrogen phosphate
    Grade Technical Grade

    Specifications

    Formula Weight 132.06
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Mesh Size −100 Mesh
    MDL Number MFCD00010891
    CAS 7783-28-0
    Merck Index 14,542
    Color White
    Melting Point 155°C (dec.)
    Quantity 1kg
    Physical Form Powder
    Synonym Diammonium hydrogen phosphate; Phosphoric acid diammonium salt
    Health Hazard 1 Warning
    Concentration P2O5 53% min.
    Density 1.619g/mL
    TSCA TSCA
    Solubility Information Freely soluble in water; Insoluble in alcohol, acetone
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    RTECS Number TB9385000
    Chemical Name or Material Ammonium hydrogen phosphate
    Grade Technical Grade

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit