Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium hydrogen sulfate, 99%, for analysis

Catalog No. p-3586390
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Glass bottle

CAS: 7803-63-6 | H5NO4S | 115.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7803-63-6
Molecular Formula H5NO4S
Molecular Weight (g/mol) 115.10
MDL Number MFCD00051073
InChI Key BIGPRXCJEDHCLP-UHFFFAOYSA-N
Synonym ammonium bisulfate, ammonium hydrogensulphate, sulfuric acid, monoammonium salt, unii-6218r7mbzb, sulfuric acid, ammonium salt 1:1, azanium hydrogen sulfate, ammonia sulfate, acmc-1bcew, ammonium hydrogen sulphate, nh4hso4
PubChem CID 24655
SMILES [NH4+].OS([O-])(=O)=O

Specifications

Boiling Point 350.0°C
Melting Point 147.0°C
Color White
Physical Form Crystalline Powder
Quantity 250 g
Assay Percent Range 99%
Linear Formula NH4HSO4
Merck Index 15, 497
Solubility Information Solubility in water: 1000g/L (20°C). Other solubilities: practically insoluble in alcohol, acetone and, pyridine
IUPAC Name azanium;hydrogen sulfate
Formula Weight 115.11
Percent Purity 99%
Grade Analytical
Packaging Glass bottle
Residue after Ignition 0.01% max. (600°C)
Chemical Name or Material Ammonium hydrogen sulfate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
EINECSNumber 232-265-5

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.