Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Hydroxide, 10% (v/v), Ricca Chemical
SDP

Catalog No. 63151 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
6315-1 4 L Natural Poly Bottle
6315-16 500 mL Natural Poly Bottle
6315-32 1 L Natural Poly Bottle
R06315004F 4 L Cubitainer
6315-5 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 6315-1 Supplier Ricca Chemical Company Supplier No. 631.51
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1336-21-6 , 7732-18-5
Molecular Formula H5NO
Molecular Weight (g/mol) 35.05
MDL Number MFCD00066650
InChI Key VHUUQVKOLVNVRT-UHFFFAOYSA-N
PubChem CID 14923
ChEBI CHEBI:18219
IUPAC Name amine hydrate
SMILES N.O

Specifications

Color Colorless
CAS Min % 90.0, 9.0
CAS Max % 94.0, 11.0
Concentration or Composition Notes 10% (v/v)
Synonym ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh
Quantity 4 L
Percent Purity 9.7 to 10.3% v/v
Grade Laboratory
Packaging Natural Poly Bottle
Solvent or Matrix Aqueous Solution
Physical Form Liquid
Chemical Name or Material Ammonium Hydroxide
Show More Show Less
ShelfLife 24 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.