Learn More
Specifications
Specifications
| Color | Colorless |
| CAS Min % | 90.0, 9.0 |
| CAS Max % | 94.0, 11.0 |
| Concentration or Composition Notes | 10% (v/v) |
| Synonym | ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh |
| Quantity | 20 L |
| Percent Purity | 9.7 to 10.3% v/v |
| Grade | Laboratory |
| Packaging | Cubitainer |
| Solvent or Matrix | Aqueous Solution |
| Show More |
Safety and Handling
| ShelfLife | 24 months |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
