Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Hydroxide, 28.0% to 30.0%, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Catalog No. 02002292
Click to view available options
Quantity:
2.5 L
4 L
Packaging:
Clear Glass Bottle
Pressure Bottom Poly Bottle

High quality chemicals for laboratory and specialized industrial use

Chemical Identifiers

CAS 1336-21-6
Molecular Formula H5NO
Molecular Weight (g/mol) 35.05
MDL Number MFCD00066650
InChI Key VHUUQVKOLVNVRT-UHFFFAOYSA-N
Synonym ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh
PubChem CID 14923
ChEBI CHEBI:18219
IUPAC Name amine hydrate
SMILES N.O

Specifications

Quantity 4 L
Formula Weight 35.05
Percent Purity 28.0 to 30.0%
Grade BAKER ANALYZED™ ACS Reagent
Packaging Pressure Bottom Poly Bottle
Specific Gravity 1L = 0.90kg
Chemical Name or Material Ammonium Hydroxide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.