Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Hydroxide 29%, CMOS, J.T.Baker™ Non-distribution product as customer accommodation.

Click to view available options
Quantity:
4 x 8PT
1336-21-6
H5NO
35.05
MFCD00066650
VHUUQVKOLVNVRT-UHFFFAOYSA-N
ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh
14923
CHEBI:18219
ammonium hydroxide
[NH4+].[OH-]
1336-21-6
H5NO
35.05
MFCD00066650
VHUUQVKOLVNVRT-UHFFFAOYSA-N
ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh
14923
CHEBI:18219
ammonium hydroxide
[NH4+].[OH-]
Assay Percent Range 29%
Quantity 4 x 8PT
Chemical Name or Material Ammonium Hydroxide