Ammonium hydroxide, 5.0N Standardized Solution, Alfa Aesar™

Manufacturer: Alfa Aesar 35577K2

Quantity 1L

 View more versions of this product

Catalog No. AA35577K2

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1336-21-6
    Physical Form Liquid
    Molecular Formula H5NO
    Formula Weight 35.05
    MDL Number MFCD00066650
    Chemical Name or Material Ammonium hydroxide

    Specifications

    Formula Weight 35.05
    Quantity 1L
    Color Colorless
    DOT Information DOT Class: 8; Packing Group: III
    EINECS Number 215-647-6
    Physical Form Liquid
    Molecular Formula H5NO
    Appearance Colorless liquid
    MDL Number MFCD00066650
    CAS 1336-21-6
    Health Hazard 1 H314-H400-H302-H335: Causes severe skin burns and eye damage. Very toxic to aquatic life. Harmful if swallowed. May cause respiratory irritation.
    Chemical Name or Material Ammonium hydroxide

    H314-H400-H302-H335: Causes severe skin burns and eye damage. Very toxic to aquatic life. Harmful if swallowed. May cause respiratory irritation.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit