Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Hydroxide, 5% (v/v), Ricca Chemical
SDP

Catalog No. 62832 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
628-32 1 L Natural Poly Bottle
628-16 500 mL Natural Poly Bottle
628-1 4 L Natural Poly Bottle
628-5 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 628-32 Supplier Ricca Chemical Company Supplier No. 62832
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1336-21-6

Specifications

Quantity 1 L
Percent Purity 4.9 to 5.1%
Grade Laboratory
Packaging Natural Poly Bottle
Chemical Name or Material Ammonium Hydroxide Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.