Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Hydroxide, 6.0 N Solution, Spectrum™ Chemical
SDP

Catalog No. 18600604
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Poly Bottle

A-540, 1336-21-6

Spectrum™ Chemical Ammonium Hydroxide, 6.0 N Solution , is a solution of ammonia in water. Aqueous ammonia, as it′s otherwise known is used as a complexant and base in qualitative inorganic analysis. It becomes a deep blue color with copper solutions. Used to dissolve silver oxide residues, such as the ones formed from Tollens′ reagent, it is also commonly used in numerous cleaning agents, such as window cleaning formulas as well as a complexant that is manufactured to parts-per-billion trace impurities and has been used in the semiconductor industry to dissolve heavy or precious metal components.

Chemical Identifiers

CAS 1336-21-6, 7732-18-5
Molecular Formula H5NO
Molecular Weight (g/mol) 35.05
MDL Number MFCD00066650
InChI Key VHUUQVKOLVNVRT-UHFFFAOYSA-N
IUPAC Name amine hydrate
SMILES N.O

Specifications

CAS Max % 6%, 92%
Quantity 500 mL
UN Number UN2672
Concentration or Composition (by Analyte or Components) 6.0 N +/-0.2
Packaging Poly Bottle
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.