Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ammonium Hydroxide, ACS Reagent (Aqua Ammonia), Reagents
Click to view available options
Quantity:
500 mL
2.5 L
20 L
194 L
Packaging:
Glass Bottle
HDPE Winpak
Poly Drum
Safety Coated Glass
Specifications
Specifications
pH | 13.6 |
Color | Colorless |
Boiling Point | 36°C |
Quantity | 20 L |
Grade | ACS |
Packaging | HDPE Winpak |
Specific Gravity | 0.9 |
Physical Form | Liquid |
Chemical Name or Material | Ammonium Hydroxide, ACS Reagent (Aqua Ammonia) |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction