Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Hydroxide, ACS Reagent (Aqua Ammonia), Reagents

Catalog No. 01336792
Change view
Click to view available options
Quantity:
500 mL
2.5 L
Packaging:
Glass Bottle
Safety Coated Glass
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
01-336-792 2.5 L Glass Bottle
01-336-793 2.5 L Safety Coated Glass
01-336-790 500 mL Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-336-792 Supplier Reagents Holdings Llc Supplier No. C51035002.5D1
Add to Cart
Add to Cart

Ammonium Hydroxide, ACS Reagent (Aqua Ammonia), ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 1336-21-6
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Aqua Ammonia
IUPAC Name water
SMILES O

Specifications

pH 13.6
Color Colorless
Boiling Point 36°C
Quantity 2.5 L
Grade ACS
Packaging Glass Bottle
Specific Gravity 0.9
Physical Form Liquid
Chemical Name or Material Ammonium Hydroxide, ACS Reagent (Aqua Ammonia)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.