Learn More
Ammonium Hydroxide Solution, ACS Reagent, 28.0 to 30.0% NH3 basis, Solstice
Shop All Honeywell Research Chemicals ProductsDescription
- Leading the Industry For Over 65 Years
- Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| Linear Formula | NH4OH |
| Concentration | 28 to 30% NH3 basis |
| Beilstein | 3587154 |
| Density | 0.9g/mL at 25°C |
| Quantity | 1 L |
| IUPAC Name | azanium;hydroxide |
| Explosion Limit | 0.27 |
| Formula Weight | 35.05g/mol |
| Grade | ACS Reagent |
| Packaging | Glass Bottle |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.