Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium hydroxide solution, Puriss. p.a., Reag. ISO, Reag. Ph. Eur., ∼25% NH3 basis, Solstice

Catalog No. 60045823 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
5 L
4 x 2.5 L
4 x 5 L
6 x 1 L
Packaging:
Plastic Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-045-823 4 x 2.5 L Plastic Bottle
60-003-24 5 L Plastic Bottle
60-045-826 6 x 1 L Plastic Bottle
60-045-825 4 x 5 L Plastic Bottle
60-003-20 1 L Plastic Bottle
60-003-22 2.5 L Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 60-045-823 Supplier Solstice Supplier No. 305014X2.5L
Add to Cart
Add to Cart

Puriss. p.a., Reag. ISO, Reag. Ph. Eur., ∼25% NH3 basis

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1336-21-6 , 7732-18-5
Molecular Formula H5NO
Molecular Weight (g/mol) 35.05
MDL Number MFCD00066650
InChI Key VHUUQVKOLVNVRT-UHFFFAOYSA-N
Synonym ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh
PubChem CID 14923
ChEBI CHEBI:18219
IUPAC Name azanium;hydroxide
SMILES N.O

Specifications

Boiling Point 24°C
CAS Min % ≥70.0000%, ≥20.0000%
CAS Max % <90.0000%, <30.0000%
Linear Formula NH4OH
UN Number UN2672
Beilstein 3587154
Density 0.892-0.910 g/mL at 20 °C; 0.9 g/mL at 25 °C(lit.)
Quantity 4 x 2.5 L
Percent Purity >25%
Grade puriss. p.a.
Packaging Plastic Bottle
Chemical Name or Material Ammonium hydroxide solution, Puriss. p.a., Reag. ISO, Reag. Ph. Eur., ∼25% NH3 basis
Show More Show Less
ShelfLife 1440 days from date of manufacture
DOTInformation Transport Hazard Class: 8; Packing Group:3: III; Proper Shipping Name: Ammonia solutions
Recommended Storage Room Temp
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.