Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium iodate, 99%

Catalog No. p-7049496
Click to view available options
Quantity:
50 g
250 g

CAS: 13446-09-8 | H4INO3 | 192.94 g/mol

Ammonium iodate is used as an oxidation reagent. Ammonium Iodate-18O3 is the labeled analog of ammonium iodate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13446-09-8
Molecular Formula H4INO3
Molecular Weight (g/mol) 192.94
MDL Number MFCD00049684
InChI Key ZRDJERPXCFOFCP-UHFFFAOYSA-N
Synonym ammonium iodate, azanium iodate, iodic acid hio3 , ammonium salt, acmc-1bqbr, ammonium iodate 50g, iodic acid hio3 , ammonium salt 1:1
PubChem CID 166805
IUPAC Name azanium;iodate
SMILES [NH4+].[O-]I(=O)=O

Specifications

Melting Point 150°C
Color White
Physical Form Crystalline
Quantity 50 g
Linear Formula NH4IO3
UN Number UN1479
Solubility Information Soluble in water 20.6g/L @ 20°C
Formula Weight 192.94
Percent Purity 99%
Odor Odorless
Sensitivity Moisture sensitive
Density 3.309 g/mL
Chemical Name or Material Ammonium iodate
Show More Show Less
Hazard Category H272-H302-H315-H319-H335
Hazard Statement GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P220-P221-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: OXIDIZING SOLID, N.O.S.
EINECSNumber 236-592-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.