Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium iodide, 99.995% (metals basis)

Catalog No. AA4025514
Click to view available options
Quantity:
5 g
25 g
100 g

Light Sensitive and Hygroscopic | CAS: 12027-06-4 | H4IN | 144.94 g/mol

It is used in photographic chemicals and some medications. Ammonium iodide and hydrogen peroxide plays an essential role for the eco-friendly oxyiodination of aromatic compounds. Ammonium Iodide and DMF is also useful for the copper-mediated selective cyanation reaction of Indoles and 2-Phenylpyridines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12027-06-4
Molecular Formula H4IN
Molecular Weight (g/mol) 144.94
MDL Number MFCD00011424
InChI Key UKFWSNCTAHXBQN-UHFFFAOYSA-N
Synonym ammonium iodide, azanium iodide, unii-oz8f027ldh, ammonium iodide nh4 i, oz8f027ldh, ammonium-iodid, ammonium jodatum, ammoniumiodid, hsdb 476, nh4i
PubChem CID 25487
ChEBI CHEBI:63040
IUPAC Name azanium;iodide
SMILES [NH4+].[I-]

Specifications

Melting Point 551° (sublimation)
Color White
Physical Form Granules
Quantity 25 g
Assay Percent Range 99.995% (Metals basis)
Solubility Information Soluble in water and ethanol.
Formula Weight 144.94
Odor Odorless
Refractive Index 1.7031
Sensitivity Air and light sensitive; Hygroscopic
Appearance White granules
Vapor Pressure 1.3 hPa (1mm Hg)
Density 2.514 g/cm3
Chemical Name or Material Ammonium Iodide
Show More Show Less
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 234-717-7
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.