Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium iodide, 99%, extra pure

Catalog No. p-4813317
Click to view available options
Quantity:
100 g
500 g
2.5 kg

Ammonium iodide, 99%, H4IN, CAS Number-12027-06-4, unii-oz8f027ldh, ammonium iodide, hsdb 476, ammoniumiodid, ammonium iodide nh4 i, nh4i, oz8f027ldh, ammonium jodatum, azanium iodide, ammonium-iodid, 500g, 98.5% min. (Argentometry), CHEBI:63040, 2.5100g/mL, 234-717-7, 144.94, Extra Pure | CAS: 12027-06-4 | H4IN | 144.94 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12027-06-4
Molecular Formula H4IN
Molecular Weight (g/mol) 144.94
MDL Number MFCD00011424
InChI Key UKFWSNCTAHXBQN-UHFFFAOYSA-N
Synonym ammonium iodide, azanium iodide, unii-oz8f027ldh, ammonium iodide nh4 i, oz8f027ldh, ammonium-iodid, ammonium jodatum, ammoniumiodid, hsdb 476, nh4i
PubChem CID 25487
ChEBI CHEBI:63040
IUPAC Name azanium;iodide
SMILES [NH4+].[I-]

Specifications

Melting Point 551.0°C
Quantity 100 g
Assay Percent Range 98.5% min. (Argentometry)
Linear Formula NH4I
Merck Index 15,526
Solubility Information Solubility in water: 1770g/L (25°C). Other solubilities: 2500g/L (100°C), 270g/L alcohol - 667g/L glycerol, 400g/L methanol
Formula Weight 144.94
Percent Purity 99%
Grade Extra Pure
Packaging Glass bottle
Residue after Ignition 0.05% max
Specific Gravity 2.51
Density 2.5100g/mL
Chemical Name or Material Ammonium iodide
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Avoid breath
EINECSNumber 234-717-7

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.