Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Iodide, ACS Reagent, Reagents

Catalog No. 01338101
Change view
Click to view available options
:
Quantity: 100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01-338-101 Quantity: 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-338-101 Supplier Reagents Holdings Llc Supplier No. C1134500100C2
Only null left
Add to Cart
Add to Cart

Ammonium Iodide, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 12027-06-4
Molecular Formula H4IN
Molecular Weight (g/mol) 144.94
InChI Key UKFWSNCTAHXBQN-UHFFFAOYSA-N
IUPAC Name ammonium iodide
SMILES [NH4+].[I-]

Specifications

Boiling Point 405°C
Melting Point 551°C
Color Colorless/White to Pale Yellow
Physical Form Solid
Quantity 100 g
Grade ACS
Packaging Glass Bottle
Specific Gravity 2.514
Chemical Name or Material Ammonium Iodide, ACS Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.