Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Potassium iodide, Puriss., meets analytical spec. of Ph. Eur. BP, USP, 99.0-100.5% (calc. to the dried substance), Honeywell Fluka™
Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
250 g
1 kg
5 kg
6 x 250 g
6 x 1 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Specifications
Specifications
Boiling Point | 1,323°C |
CAS Max % | 100.0000% |
Quantity | 6 x 1 kg |
Assay Percent Range | 99.0 to 100.5% |
Linear Formula | KI |
Formula Weight | 166 |
Grade | puriss. |
Loss on Drying | max. 0.5% |
Packaging | Poly Bottle |
Density | 3.12 g/cm3 |
Show More |
Safety and Handling
ShelfLife | 360 days from date of manufacture |
EINECSNumber | 231-659-4 |
RTECSNumber | TT2975000 |
Recommended Storage | Room Temp |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction