Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium L-(+)-tartrate, 98%

Catalog No. AA1765830
Click to view available options
Quantity:
50 g
250 g
1 kg

CAS: 3164-29-2 | C4H12N2O6 | 184.15 g/mol

Ammonium L-(+)-tartrate finds application in the textile and pharmaceutical industries. It is also used to study protein structural analysis, biochemicals and reagents, biological buffers, chelators and proteomics. It acts as a nitrogen source in biological applications and cell culture media. Further, it is used in the study of crystal optimization research and preliminary diffraction data analysis of the Smad1 MH1 domain bonded to a palindromic SBE DNA element.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3164-29-2
Molecular Formula C4H12N2O6
Molecular Weight (g/mol) 184.15
MDL Number MFCD00013073
InChI Key NGPGDYLVALNKEG-OLXYHTOASA-N
Synonym ammonium d-tartrate, diammonium l-+-tartrate, l-tartaric acid, ammonium salt, unii-ji8ud88354, diammonium l-tartrate, r-2,3-dihydroxybutanedioic acid diammonium salt, ammonium tartrate dibasic, butanedioic acid, 2,3-dihydroxy-2r,3r-, diammonium salt, ammonium l-+-tartrate, diammonium 2r,3r-2,3-dihydroxysuccinate
PubChem CID 13652278
ChEBI CHEBI:63075
IUPAC Name (2R,3R)-2,3-dihydroxybutanedioic acid diamine
SMILES N.N.O[C@H]([C@@H](O)C(O)=O)C(O)=O

Specifications

Physical Form Crystalline
Quantity 250 g
Beilstein 6120352
Solubility Information Soluble in water and alcohol.
Formula Weight 184.15
Sensitivity Hygroscopic
Density 1.6
Chemical Name or Material Ammonium-L-tartrate
EINECSNumber 221-618-9
RTECSNumber WW8050000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.