Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ammonium metavanadate, 99.5%, for analysis
Click to view available options
Quantity:
50 g
250 g
1 kg
5 kg
Packaging:
Glass bottle
Plastic Drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 200.0°C |
Color | Green to White |
Physical Form | Crystalline Powder |
Quantity | 250 g |
Assay Percent Range | 99.5% |
Linear Formula | NH4VO3 |
Fieser | 01,1057 |
Merck Index | 15,565 |
Solubility Information | Solubility in water: 5.1g/L (20°C). Other solubilities: insoluble in ethanol, 69.5g/L (96°C) |
IUPAC Name | azanium;oxido(dioxo)vanadium |
Show More |
Safety and Handling
Hazard Category | GHS Signal Word: Danger |
Hazard Statement | GHS H Statement Suspected of damaging the unborn child. Causes damage to organs through prolonged or repeated exposure if inhaled. Toxic if swallowed. Causes serious eye irritation. Harmful if inhaled. Toxic to aquati |
Precautionary Statement | GHS P Statement IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician if you feel unwell. |
EINECSNumber | 232-261-3 |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction