Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium metavanadate, 99.5%, for analysis

Catalog No. p-4509711
Click to view available options
Quantity:
50 g
250 g
1 kg
5 kg
Packaging:
Glass bottle
Plastic Drum

CAS: 7803-55-6 | H4NO3V | 116.98 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7803-55-6
Molecular Formula H4NO3V
Molecular Weight (g/mol) 116.98
MDL Number MFCD00011430
InChI Key YBVKNHXQSRDWAA-UHFFFAOYSA-M
Synonym ammonium metavanadate, ammonium vanadate v, ammonium monovanadate, unii-fl85px638g, vanadate vo31-, ammonium, vanadate vo31-, ammonium 1:1, ammonium vanadium oxide, ammoniummetavanadate, vanadic acid, hvo3 , ammonium salt, ammonium meta-vanadate
PubChem CID 516859
SMILES N.O[V](=O)=O

Specifications

Melting Point 200.0°C
Color Green to White
Physical Form Crystalline Powder
Quantity 50 g
Assay Percent Range 99.5%
Linear Formula NH4VO3
Fieser 01,1057
Merck Index 15,565
Solubility Information Solubility in water: 5.1g/L (20°C). Other solubilities: insoluble in ethanol, 69.5g/L (96°C)
IUPAC Name azanium;oxido(dioxo)vanadium
Formula Weight 116.98
Percent Purity 99.5%
Grade Analytical
Packaging Glass bottle
Chemical Name or Material Ammonium metavanadate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Suspected of damaging the unborn child.
Causes damage to organs through prolonged or repeated exposure if inhaled.
Toxic if swallowed.
Causes serious eye irritation.
Harmful if inhaled.
Toxic to aquati
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unwell.
EINECSNumber 232-261-3

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.