Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Imidazo[1,2-b]pyridazine, 98%, Thermo Scientific™

Catalog No. AC455660050
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 766-55-2
Molecular Formula C6H5N3
Molecular Weight (g/mol) 119.12
InChI Key VTVRXITWWZGKHV-UHFFFAOYSA-N
Synonym imidazo 1,2-b pyridazine, imidazol 1,2-b pyridazine, imdazo1,2-bpyridazine, pubchem11271, acmc-1bczx, imidazo 2,1-f pyridazine, 1,5-diazaindolizine, ksc258e5b, 1,3a,4-triazaindene
PubChem CID 136599
IUPAC Name imidazo[1,2-b]pyridazine
SMILES C1=CC2=NC=CN2N=C1

Specifications

Melting Point 54.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Quantity 5 g
Solubility Information Solubility in water: .. Other solubilities: soluble in dmf
Formula Weight 119.12
Percent Purity 98%
Chemical Name or Material Imidazo[1,2-b]pyridazine
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.