Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium molybdate, 99.99%, (trace metal basis)

Catalog No. p-4813322
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle

CAS: 13106-76-8 | H8MoN2O4 | 196.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13106-76-8
Molecular Formula H8MoN2O4
Molecular Weight (g/mol) 196.01

Specifications

Quantity 5 g
Assay Percent Range 99.99%
Linear Formula (NH4)2MoO4
Solubility Information Solubility in water: soluble. Other solubilities: practically insoluble in alcohol
Formula Weight 196.01
Percent Purity 99.99%
Grade Trace Metal Basis
Packaging Glass bottle
Chemical Name or Material Ammonium molybdate
Hazard Category GHS Signal Word: Warning
EINECSNumber 236-031-3

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.