Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium molybdate (di), 4% w/v aq. soln.

Catalog No. p-7050093
Click to view available options
Quantity:
500 mL
1 L

CAS: 27546-07-2 | H2MoO4 | 161.96 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27546-07-2
Molecular Formula H2MoO4
Molecular Weight (g/mol) 161.96
MDL Number MFCD00010882
InChI Key VLAPMBHFAWRUQP-UHFFFAOYSA-L

Specifications

Concentration 4% w/v aq. soln.
Formula Weight 339.95
Quantity 500 mL
Physical Form Liquid
Chemical Name or Material Ammonium molybdate (di)
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.