Alfa Aesar™ Ammonium molybdate (di), Mo 56.5%

Manufacturer: Alfa Aesar™ 12215A1

Quantity 1kg

 View more versions of this product

Catalog No. AA12215A1

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 27546-07-2
    Formula Weight 339.95
    MDL Number MFCD00010882
    Synonym Ammonium dimolybdate
    Chemical Name or Material Ammonium molybdate (di)

    Specifications

    Formula Weight 339.95
    MDL Number MFCD00010882
    TSCA TSCA
    Physical Form Powder
    CAS 27546-07-2
    Synonym Ammonium dimolybdate
    Quantity 1kg
    Concentration Mo 56.5%
    Chemical Name or Material Ammonium molybdate (di)

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit